Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QSSYSSYGQS QNTGYGTQST PQGYGSTGGY GSSQSSQSSY GQQSSYPGYG QQPAPSSTSG SYGSSSQSSS YGQPQSGSYS QQPSYGGQQQ SYGQQQSYNP PQGYGQQNQY NSSSGGGGGG GGGGNYGQDQ SSMSSGGGSG GGYGNQDQSG GGGSGGYGQQ DRGGRGRGGS GGGGGGGGGG YNRSSGGYEP RGRGGGRGGR GGMGGSDRGG FNKFGGPRDQ GSRHDSEQDN SDNNTIFVQG LGENVTIESV ADYFKQIGII KTNKKTGQPM INLYTDRETG KLKGEATVSF DDPPSAKAAI DWFDGKEFSG NPIKVSFATR RADFNRGGGN GRGGRGRGGP MGRGGYGGG
This gene encodes a multifunctional protein component of the heterogeneous nuclear ribonucleoprotein (hnRNP) complex. The hnRNP complex is involved in pre-mRNA splicing and the export of fully processed mRNA to the cytoplasm. This protein belongs to the FET family of RNA-binding proteins which have been implicated in cellular processes that include regulation of gene expression, maintenance of genomic integrity and mRNA/microRNA processing. Alternative splicing results in multiple transcript variants. Defects in this gene result in amyotrophic lateral sclerosis type 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 16) in malignant liposarcoma); 16) malignant liposarcoma; 75 kDa DNA-pairing protein; fus-like protein; fused in sarcoma; fusion (involved in t(12; fusion gene in myxoid liposarcoma; fusion, derived from t(12; fusion, derived from t(12;16) malignant liposarcoma; heterogeneous nuclear ribonucleoprotein P2; hnRNP P2; Oncogene FUS; Oncogene TLS; pigpen protein; POMp75; Protein pigpen; RNA-binding protein FUS; translocated in liposarcoma; Translocated in liposarcoma protein
Gene Aliases: ALS6; D430004D17Rik; D930039C12Rik; ETM4; FUS; FUS1; HNRNPP2; POMP75; TLS
UniProt ID: (Human) P35637, (Mouse) P56959
Entrez Gene ID: (Human) 2521, (Rat) 317385, (Mouse) 233908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support