Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: YSGTWNGNTK VAIKTLKPGT MSPESFLEEA QIMKKLKHDK LVQLYAVVSE EPIYIVTEYM NKGSLLDFLK DGEGRALKLP NLVDMAAQVA AGMAYIERMN YIHRDLRSAN ILVGNGLICK IADFGLARLI EDNEYTARQG AKFPIKWTAP EAALYGRFTI KSDVWSFGIL LTELVTKGRV PYPGMNNREV LEQVERGYRM PCPQDCPISL HELMIHCWKK DPEERPTFEY LQSFLEDYFT ATEPQYQPGE NL
FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; FYN oncogene related to SRC, FGR, YES; fyn {ECO:0000312|RGD:2641}; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; Protooncogene Syn; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; Src-like kinase; src/yes-related novel; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn
Gene Aliases: AI448320; AW552119; FYN; p59-FYN; SLK; SYN
UniProt ID: (Human) P06241, (Rat) Q62844, (Mouse) P39688
Entrez Gene ID: (Human) 2534, (Rat) 25150, (Mouse) 14360
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support