Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 15 µL or 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABA-B receptor 1; GABA-B receptor, R1 subunit; GABA-B-R1; Gaba-b-r1a; Gaba-b1; GABA-BR1; Gaba-br1a receptor; GABA-BR1b receptor; GABAB; GABABR1; Gababr1 receptor; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor, 1; gamma-aminobutyric acid (GABA-B) receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; gb1; GBR1; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor
Gene Aliases: bM573K1.1; GABAB1; GABABR1; GABBR1; GABBR1-3; GB1; GPRC3A
UniProt ID: (Human) Q9UBS5, (Mouse) Q9WV18, (Rat) Q9Z0U4
Entrez Gene ID: (Human) 2550, (Mouse) 54393, (Rat) 81657
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support