Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAK serine/threonine kinase or Cyclin-G-associated kinase was originally identified as a protein that is associated with cyclin G. However, neither CDK5 nor GAK kinase activity is activated by cyclin G nor does GAK affect CDK5 kinase activity. GAK seems to act as an auxilin homolog that is involved in the uncoating of clathrin-coated vesicles by Hsc70 in non-neuronal cells. Expression oscillates slightly during the cell cycle, peaking at G1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: auxilin-2; cyclin G-associated kinase; Cyclin-G-associated kinase; DnaJ homolog subfamily C member 26
Gene Aliases: D130045N16Rik; DNAJ26; DNAJC26; GAK
UniProt ID: (Human) O14976, (Mouse) Q99KY4, (Rat) P97874
Entrez Gene ID: (Human) 2580, (Mouse) 231580, (Rat) 81659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support