Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NYFLCGLKG IQEHFGLSNL TGMNCLVDGN IPPSSGLSSS SALVCCAGLV TLTVLGRNLS KVELAEICAK SERYIGTEGG GMDQSISFLA EEGTAKLIEF SPLRATDVKL PSGAVFVIAN SCVEMNKAAT SHFNIRVMEC RLAAKLLAKY KSLQWDKVLR LEEVQAKLGI SLEEMLLVTE DALHPEPYNP EEICRCLGIS LEELRTQILS PNTQDVLIFK LYQRAKHVYS EAARVLQFKK ICEEAPENMV QLLGELMNQS HMSCRDMYEC SCPELDQLVD ICRKFGAQGS RLTGAGWGGC TVSMVPADKL PSFLANVHKA YYQRSDGSLA PEKQSLFATK PGGGALVLLE A (109-458 aa encoded by BC005141)
This gene encodes a highly efficient N-acetylgalactosamine kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Galactokinase 2; GalNAc kinase; N-acetylgalactosamine kinase
Gene Aliases: 2810017M24Rik; AI327030; GALK2; GK2
UniProt ID: (Human) Q01415, (Mouse) Q68FH4, (Rat) Q5XIG6
Entrez Gene ID: (Human) 2585, (Mouse) 69976, (Rat) 296117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support