Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLLDENVCLD QGPFPGNTPI MYYCHEFSSQ NVYYHLTGEL YVGQLIAEAS ASDRCLTDPG KAEKPTLEPC SKAAKNRLHI YWDFKPGGAV INRDTKRCLE MKKDLLGSHV LVLQTCSTQV WEIQHTVR
Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 41%.
Probably catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc transferase 8; GalNAc-T8; Polypeptide GalNAc transferase 8; pp-GaNTase 8; Probable polypeptide N-acetylgalactosaminyltransferase 8; Protein-UDP acetylgalactosaminyltransferase 8; UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase 8; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 8; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 8 (GalNAc-T8)
Gene Aliases: GALNAC-T8; GALNT8
UniProt ID: (Human) Q9NY28
Entrez Gene ID: (Human) 26290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support