Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQHRFVFKR CGPDGQWVRG PRGQPWRDAS QCQMDGEEIE VQKEVAKMYS SF (91-141 aa encoded by BC104854)
Glucagon receptors are G-protein coupled receptors for glucagon and play a central role in the regulation of blood glucose levels and glucose homeostatsis. It regulates the rate of hepatic glucose production by promoting glycogen hydrolysis and gluconeogenesis. Glucagon receptors also play an important role in mediating the responses to fasting. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G-proteins) and modulates the activity of down-stream effectors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GL-R; Glucagon receptor
Gene Aliases: GCGR; GGR; GL-R; GR
UniProt ID: (Human) P47871, (Mouse) Q61606
Entrez Gene ID: (Human) 2642, (Mouse) 14527
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support