Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDP-D-glucose phosphorylase 1; GDP-D-glucose phosphorylase C15orf58; GDP-D-glucose phosphorylase C15orf58 homolog
Gene Aliases: C15orf58; D330012F22Rik; GDPGP1; RGD1560277; VTC2
UniProt ID: (Human) Q6ZNW5, (Mouse) Q3TLS3
Entrez Gene ID: (Human) 390637, (Mouse) 269952, (Rat) 308763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support