Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FNIISPSNN GGNVQETVTI DNEKNTAIIN IHAGSCSSTT IFDYKHGYIA SRVLSRRACF ILKMDHQNIP PLNNLQWYIY EKQALDNMFS SKYTWVKYNP LESLIKDVDW FLLGSPIEKL CKHIPLYKGE VVENTHNVGA GGCAKAGLLG ILGISICADI HV (24-184 aa encoded by BC146484)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Blottin; BRICHOS domain containing 1B; Down-regulated in gastric cancer; down-regulated in gastric cancer GDDR; Gastrokine-2; Trefoil factor interactions(z) 1
Gene Aliases: 1810036H07Rik; BLOT; BRICD1B; GDDR; GKN2; PRO813; TFIZ1; UNQ465/PRO813; VLTI465
UniProt ID: (Human) Q86XP6, (Mouse) Q9CQS6
Entrez Gene ID: (Human) 200504, (Mouse) 66284
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support