Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed in kidney and amygdala. Not expressed in other tissues or brain regions tested.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 102; G-protein coupled receptor 102; Gm1149; mTaar8a; Q923; Ta10; Ta11; Ta7; Taar8b; Taar8c; TaR-11; TaR-5; TaR-8; TaR-8a; Trace amine receptor 11; Trace amine receptor 5; trace amine receptor 8; trace amine receptor 8a; Trace amine-associated receptor 8; Trace amine-associated receptor 8a; trace-amine-associated receptor 8a
Gene Aliases: Gm230; GPR102; Ta11; TA5; TAAR8; Taar8a; TaR-5; TaR-8; taR-8a; Tar11; TAR5; Trar11; TRAR5
UniProt ID: (Human) Q969N4, (Mouse) Q5QD07, (Rat) Q923X9
Entrez Gene ID: (Human) 83551, (Mouse) 215859, (Rat) 319104
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support