Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MESVALYSF QATESDELAF NKGDTLKILN MEDDQNWYKA ELRGVEGFIP KNYIRVKPHP WYSGRISRQL AEEILMKRNH LGAFLIRESE SSPGEFSVSV NYGDQVQHFK VLREASGKYF LWEEKFNSLN ELVDFYRTTT IAKKRQIFLR DEEPLLKSPG ACFAQAQFDF SAQDPSQLSF RRGDIIEVLE RPDPHWWRGR SCGRVGFFPR SYVQPVHL (1-217 aa encoded by BC063035)
This gene encodes a member of the GRB2/Sem5/Drk family. This member functions as a cytoplasmic signaling protein which contains an SH2 domain flanked by two SH3 domains. The SH2 domain interacts with ligand-activated receptors for stem cell factor and erythropoietin, and facilitates the formation of a stable complex with the BCR-ABL oncoprotein. This protein also associates with the Ras guanine nucleotide exchange factor SOS1 (son of sevenless homolog 1) through its N-terminal SH3 domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GRB2-related adapter protein; growth factor receptor-bound protein 2-related adaptor protein; MGC64880
Gene Aliases: 8430435N19Rik; BB233594; GRAP
UniProt ID: (Human) Q13588, (Mouse) Q9CX99
Entrez Gene ID: (Human) 10750, (Rat) 363616, (Mouse) 71520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support