Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic transcription factor 2 34 kDa subunit; BTF2 p34; BTF2-p34; General transcription factor IIH polypeptide 3; General transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3, 34kDa; TFIIH basal transcription factor complex p34 subunit
Gene Aliases: 34kDa; 5033417D07Rik; BTF2; BTF2 p34; C730029A10; D5Ertd679e; GTF2H3; P34; TFB4; TFIIH
UniProt ID: (Human) Q13889, (Mouse) Q8VD76, (Rat) Q561R7
Entrez Gene ID: (Human) 2967, (Mouse) 209357, (Rat) 288651
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support