Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Hematological and neurological expressed 1-like protein (HN1L) is expressed in liver, kidney, prostate, testis and uterus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRAMP_1 like; Hematological and neurological expressed 1-like protein; HN1 like; HN1-like protein; Jupiter microtubule associated homolog 2
Gene Aliases: C16orf34; HN1L; JPT2; L11
UniProt ID: (Human) Q9H910
Entrez Gene ID: (Human) 90861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support