Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDVRRLKVNE LREELQRRGL DTRGLKAELA ERLQAALEAE EPDDERELDA DDEPGRPGHI NEEVETEGGS ELEGTAQPPP PGLQPHAEPG GYSGPDGHYA MDNITRQNQF YDTQVIKQEN ESGYERRPLE MEQQQAYRPE MKTEMKQGAP TSFLPPEASQ LKPDRQQFQS RKRPYEENRG RGYFEHREDR RGRSPQPPAE EDEDDFDDTL VAIDTYNCDL HFKVARDRSS GYPLTIEGFA YLWSGARASY GVRRGRVCFE MKINEEISVK HLPSTEPDPH VVRIGWSLDS CSTQLGEEPF SYGYGGTGKK STNSRFENYG DKFAENDVIG CFADFECGND VELSFTKNGK WMGIAFRIQK EALGGQALYP HVLVKNCAVE FNFGQRAEPY CSVLPGFTFI QHLPLSERIR GTVGPKSKAE CEILMMVGLP AAGKTTWAIK HAASNPSKKY NILGTNAIMD KMRVMGLRRQ RNYAGRWDVL IQQATQCLNR LIQIAARKKR NYILDQTNVY GSAQRRKMRP FEGFQRKAIV ICPTDEDLKD RTIKRTDEEG KDVPDHAVLE MKANFTLPDV GDFLDEVLFI ELQREEADKL VRQYNEEGRK AGPPPEKRFD NRGGGGFRGR GGGGGFQRYE NRGPPGGNRG GFQNRGGGSG GGGNYRGGFN RSGGGGYSQN RWGNNNRDNN NSNNRGSYNR APQQQPPPQQ PPPPQPPPQQ PPPPPSYSPA RNPPGASTYN KNSNIPGSSA NTSTPTVSSY SPPQTYPQPS YNQYQQYAQQ WNQYYQNQGQ WPPYYGNYDY GSYSGNTQGG TSTQ
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenovirus early region 1B-associated protein 5; E1B 55kDa associated protein 5; E1B-55 kDa-associated protein 5; E1B-AP5; EIB-55kDa associated protein 5; heterogeneous nuclear ribonucleoprotein U-like 1; Heterogeneous nuclear ribonucleoprotein U-like protein 1
Gene Aliases: E130317O14Rik; E1B-AP5; E1BAP5; Hnrnpul; HNRNPUL1; HNRPUL1
UniProt ID: (Human) Q9BUJ2, (Mouse) Q8VDM6
Entrez Gene ID: (Human) 11100, (Mouse) 232989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support