Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: DKIKKFLTES HDRQAKGKAI TKKKYIGIRM MSLTSSKAKE LKDRHRDFPD VISGAYIIEV IPDTPAEAGG LKENDVIISI NGQSVVSAND VSDVIKREST LNMVVRRGNE DIMITVIPEE IDP
HtrA1, a 51KDa protein, is a member of highly conserved mammalian HtrA serine protease family. It is a human homolog of bacterial HtrA and consists of a secretory signal sequence, an IGFBP-binding domain and a Kazal-type serine-protease inhibitor domain at the N-terminus and a highly conserved trypsin-like serine/protease domain and a PDZ domain at the C-terminus. It plays a major role in the regulation of cell growth. HtrA1 inhibits signaling mediated by TGF-beta family members and regulates many physiological processes. HtrA1 is up-regulated in placenta during pregnancy and plays an important role in implantation and placental physiology. It regulates the availability of IGFs by cleaving IGF-binding proteins. It acts as a tumor suppressor and is down-regulated in ovarian cancers and melanomas. Its role has been implicated in osteoarthritis suggesting a therapeutic intervention. Northern blot analysis detected high expression in placenta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High-temperature requirement A serine peptidase 1; IGFBP5-protease; insulin-like growth factor binding protein 5 protease; L56; protease serine 11; protease serine 11 (IGF binding); protease, serine, 11 (IGF binding); serine protease; Serine protease 11; Serine protease HTRA1; similar to bacterial htrA and degP
Gene Aliases: AI429470; ARMD7; CADASIL2; CARASIL; HTRA; HTRA1; L56; ORF480; PRSS11; RSPP11
UniProt ID: (Human) Q92743, (Rat) Q9QZK5, (Mouse) Q9R118
Entrez Gene ID: (Rabbit) 100008705, (Pig) 100515628, (Human) 5654, (Rat) 65164, (Mouse) 56213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support