Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 79%
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H1 histone family, member 3; histone 1, H1d; Histone H1.3; Histone H1c; Histone H1s-2
Gene Aliases: H1-3; H1.3; H1D; H1F3; H1s-2; HIST1H1D
UniProt ID: (Human) P16402
Entrez Gene ID: (Human) 3007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support