Product References
The RNA-binding protein landscapes differ between mammalian organs and cultured cells.
Nature communications
Perez-Perri JI,Ferring-Appel D,Huppertz I,Schwarzl T,Sahadevan S,Stein F,Rettel M,Galy B,Hentze MW
Wed Apr 12 00:00:00 EDT 2023
A splicing silencer in SMN2 intron 6 is critical in spinal muscular atrophy.
Human molecular genetics
Wang L,Ji Y,Chen Y,Bai J,Gao P,Feng P
Mon Mar 06 00:00:00 EST 2023
An mRNA processing pathway suppresses metastasis by governing translational control from the nucleus.
Nature cell biology
Navickas A,Asgharian H,Winkler J,Fish L,Garcia K,Markett D,Dodel M,Culbertson B,Miglani S,Joshi T,Yin K,Nguyen P,Zhang S,Stevers N,Hwang HW,Mardakheh F,Goga A,Goodarzi H
Thu Jun 01 00:00:00 EDT 2023
SULT1A1-dependent sulfonation of alkylators is a lineage-dependent vulnerability of liver cancers.
Nature cancer
Shi L,Shen W,Davis MI,Kong K,Vu P,Saha SK,Adil R,Kreuzer J,Egan R,Lee TD,Greninger P,Shrimp JH,Zhao W,Wei TY,Zhou M,Eccleston J,Sussman J,Manocha U,Weerasekara V,Kondo H,Vijay V,Wu MJ,Kearney SE,Ho J,McClanaghan J,Murchie E,Crowther GS,Patnaik S,Boxer MB,Shen M,Ting DT,Kim WY,Stanger BZ,Deshpande V,Ferrone CR,Benes CH,Haas W,Hall MD,Bardeesy N
Wed Mar 01 00:00:00 EST 2023
RNA G-quadruplex organizes stress granule assembly through DNAPTP6 in neurons.
Science advances
Asamitsu S,Yabuki Y,Matsuo K,Kawasaki M,Hirose Y,Kashiwazaki G,Chandran A,Bando T,Wang DO,Sugiyama H,Shioda N
Fri Feb 24 00:00:00 EST 2023
LncGMDS-AS1 promotes the tumorigenesis of colorectal cancer through HuR-STAT3/Wnt axis.
Cell death & disease
Ye D,Liu H,Zhao G,Chen A,Jiang Y,Hu Y,Liu D,Xie N,Liang W,Chen X,Zhang H,Li C,Wang J,Sun D,Chen W,Tan D,Wang Q,Wang H,Yu D,Wu B,Wang M,Cui S,Liu S,Zhang X
Mon Feb 27 00:00:00 EST 2023
LncGMDS-AS1 promotes the tumorigenesis of colorectal cancer through HuR-STAT3/Wnt axis.
Cell death & disease
Ye D,Liu H,Zhao G,Chen A,Jiang Y,Hu Y,Liu D,Xie N,Liang W,Chen X,Zhang H,Li C,Wang J,Sun D,Chen W,Tan D,Wang Q,Wang H,Yu D,Wu B,Wang M,Cui S,Liu S,Zhang X
Mon Feb 27 00:00:00 EST 2023
Hsa_circ_0001944 regulates apoptosis by regulating the binding of PARP1 and HuR in leukemia and malignant transformed cells induced by hydroquinone.
Environmental toxicology
Zhong B,Ling X,Meng J,Han Y,Zhang H,Liu Z,Chen J,Zhang H,Pan Z,Liu L
Wed Feb 01 00:00:00 EST 2023
Hsa_circ_0001944 regulates apoptosis by regulating the binding of PARP1 and HuR in leukemia and malignant transformed cells induced by hydroquinone.
Environmental toxicology
Zhong B,Ling X,Meng J,Han Y,Zhang H,Liu Z,Chen J,Zhang H,Pan Z,Liu L
Wed Feb 01 00:00:00 EST 2023
Long Noncoding RNA ENSG00000254693 Promotes Diabetic Kidney Disease via Interacting with HuR.
Journal of diabetes research
Yu Q,Lin J,Ma Q,Li Y,Wang Q,Chen H,Liu Y,Liu B
Tue May 03 00:00:00 EDT 2022
Long Noncoding RNA ENSG00000254693 Promotes Diabetic Kidney Disease via Interacting with HuR.
Journal of diabetes research
Yu Q,Lin J,Ma Q,Li Y,Wang Q,Chen H,Liu Y,Liu B
Tue May 03 00:00:00 EDT 2022
Danshensu methyl ester enhances autophagy to attenuate pulmonary fibrosis by targeting lncIAPF-HuR complex.
Frontiers in pharmacology
Zhu Q,Wang J,Ji Y,Luan J,Yue D,Liu W,Li H,Zhang J,Qu G,Lv C,Song X
Mon Dec 19 00:00:00 EST 2022
m6A reader YTHDC1 modulates autophagy by targeting SQSTM1 in diabetic skin.
Autophagy
Liang D,Lin WJ,Ren M,Qiu J,Yang C,Wang X,Li N,Zeng T,Sun K,You L,Yan L,Wang W
Wed Jun 01 00:00:00 EDT 2022
RNase L activation in the cytoplasm induces aberrant processing of mRNAs in the nucleus.
PLoS pathogens
Burke JM,Ripin N,Ferretti MB,St Clair LA,Worden-Sapper ER,Salgado F,Sawyer SL,Perera R,Lynch KW,Parker R
Tue Nov 01 00:00:00 EDT 2022
Inhibitory effect of calycosin on breast cancer cell progression through downregulating lncRNA HOTAIR and downstream targets: HuR and IGF2BP1.
Acta biochimica et biophysica Sinica
Li Y,Miao H,Wei W,Tian J,Chen J
Sun Dec 25 00:00:00 EST 2022
The solution structure of Dead End bound to AU-rich RNA reveals an unusual mode of tandem RRM-RNA recognition required for mRNA regulation.
Nature communications
Duszczyk MM,Wischnewski H,Kazeeva T,Arora R,Loughlin FE,von Schroetter C,Pradère U,Hall J,Ciaudo C,Allain FH
Thu Oct 06 00:00:00 EDT 2022
HuR Promotes the Progression of Gastric Cancer through Mediating CDC5L Expression.
Disease markers
Cui J,Cao N,Wang G,Wang F,Yang B,Wang J,Lv Y,Chen Y,Li F
Thu Apr 07 00:00:00 EDT 2022
Inhibition of RUNX1 blocks the differentiation of lung fibroblasts to myofibroblasts.
Journal of cellular physiology
Dubey S,Dubey PK,Umeshappa CS,Ghebre YT,Krishnamurthy P
Fri Apr 01 00:00:00 EDT 2022
Inhibition of RUNX1 blocks the differentiation of lung fibroblasts to myofibroblasts.
Journal of cellular physiology
Dubey S,Dubey PK,Umeshappa CS,Ghebre YT,Krishnamurthy P
Fri Apr 01 00:00:00 EDT 2022
Knockdown of circHIPK3 promotes the osteogenic differentiation of human bone marrow mesenchymal stem cells through activating the autophagy flux.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhuang Z,Jin C,Li X,Han Y,Yang Q,Huang Y,Zheng Y,Li W
Tue Nov 01 00:00:00 EDT 2022
Knockdown of circHIPK3 promotes the osteogenic differentiation of human bone marrow mesenchymal stem cells through activating the autophagy flux.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhuang Z,Jin C,Li X,Han Y,Yang Q,Huang Y,Zheng Y,Li W
Tue Nov 01 00:00:00 EDT 2022
Inhibition of translocator protein 18 kDa suppressed the progression of glioma via the ELAV-like RNA-binding protein 1/MAPK-activated protein kinase 3 axis.
Bioengineered
Wang J,Ren P,Zeng Z,Ma L,Li Y,Zhang H,Guo W
Tue Mar 01 00:00:00 EST 2022
CircEIF3H-IGF2BP2-HuR scaffold complex promotes TNBC progression via stabilizing HSPD1/RBM8A/G3BP1 mRNA.
Cell death discovery
Song X,Chen B,Liang Y,Li Y,Zhang H,Han D,Wang Y,Ye F,Wang L,Zhao W,Yang Q
Sat May 14 00:00:00 EDT 2022
Smooth muscle-specific HuR knockout induces defective autophagy and atherosclerosis.
Cell death & disease
Liu S,Jiang X,Cui X,Wang J,Liu S,Li H,Yang J,Zhang C,Zhang W
Fri Apr 09 00:00:00 EDT 2021
FUS/TLS Suppresses Enterovirus Replication and Promotes Antiviral Innate Immune Responses.
Journal of virology
Xue YC,Ng CS,Mohamud Y,Fung G,Liu H,Bahreyni A,Zhang J,Luo H
Mon May 24 00:00:00 EDT 2021
S-adenosylmethionine upregulates the angiotensin receptor-binding protein ATRAP via the methylation of HuR in NAFLD.
Cell death & disease
Guo T,Dai Z,You K,Battaglia-Hsu SF,Feng J,Wang F,Li B,Yang J,Li Z
Mon Mar 22 00:00:00 EDT 2021
Analysis of m6A-Related Signatures in the Tumor Immune Microenvironment and Identification of Clinical Prognostic Regulators in Adrenocortical Carcinoma.
Frontiers in immunology
Jin Y,Wang Z,He D,Zhu Y,Hu X,Gong L,Xiao M,Chen X,Cheng Y,Cao K
Thu Sep 23 00:00:00 EDT 2021
LINC00467 Promotes Tumor Progression via Regulation of the NF-kb Signal Axis in Bladder Cancer.
Frontiers in oncology
Xiao J,Gong L,Xiao M,He D,Xiang L,Wang Z,Cheng Y,Deng L,Cao K
Sat Apr 23 00:00:00 EDT 2022
Nuclear depletion of RNA-binding protein ELAVL3 (HuC) in sporadic and familial amyotrophic lateral sclerosis.
Acta neuropathologica
Diaz-Garcia S,Ko VI,Vazquez-Sanchez S,Chia R,Arogundade OA,Rodriguez MJ,Traynor BJ,Cleveland D,Ravits J
Wed Dec 01 00:00:00 EST 2021
UBE2C mRNA expression controlled by miR-300 and HuR determines its oncogenic role in gastric cancer.
Biochemical and biophysical research communications
Wang Y,Huang F,Liu M,Zhao Q
Fri Jan 01 00:00:00 EST 2021
Circular RNA circRHOBTB3 represses metastasis by regulating the HuR-mediated mRNA stability of PTBP1 in colorectal cancer.
Theranostics
Chen J,Wu Y,Luo X,Jin D,Zhou W,Ju Z,Wang D,Meng Q,Wang H,Fu X,Xu J,Song Z
Wed Jul 28 00:00:00 EDT 2021
Global analysis of RNA-binding protein dynamics by comparative and enhanced RNA interactome capture.
Nature protocols
Perez-Perri JI,Noerenberg M,Kamel W,Lenz CE,Mohammed S,Hentze MW,Castello A
Fri Jan 01 00:00:00 EST 2021
Long noncoding RNA DUXAP10 promotes the stemness of glioma cells by recruiting HuR to enhance Sox12 mRNA stability.
Environmental toxicology
Wu B,Yang C,Fang Y,Ding W,Zhang Y
Sat May 01 00:00:00 EDT 2021
Long noncoding RNA DUXAP10 promotes the stemness of glioma cells by recruiting HuR to enhance Sox12 mRNA stability.
Environmental toxicology
Wu B,Yang C,Fang Y,Ding W,Zhang Y
Sat May 01 00:00:00 EDT 2021
HuR stabilizes HTT mRNA via interacting with its exon 11 in a mutant HTT-dependent manner.
RNA biology
Zhao Q,Li C,Yu M,Sun Y,Wang J,Ma L,Sun X,Lu B
Wed Apr 01 00:00:00 EDT 2020
Cdh1-APC Regulates Protein Synthesis and Stress Granules in Neurons through an FMRP-Dependent Mechanism.
iScience
Valdez-Sinon AN,Lai A,Shi L,Lancaster CL,Gokhale A,Faundez V,Bassell GJ
Fri May 22 00:00:00 EDT 2020
MicroRNA-145 suppresses gastric cancer progression by targeting Hu-antigen R.
American journal of physiology. Cell physiology
Li Q,Tong D,Guo C,Wu F,Li F,Wang X,Jiang Q,Wei Y,Liu L,Ni L,Guo B,Huang C
Sun Mar 01 00:00:00 EST 2020
MicroRNA-145 suppresses gastric cancer progression by targeting Hu-antigen R.
American journal of physiology. Cell physiology
Li Q,Tong D,Guo C,Wu F,Li F,Wang X,Jiang Q,Wei Y,Liu L,Ni L,Guo B,Huang C
Sun Mar 01 00:00:00 EST 2020
Typical Stress Granule Proteins Interact with the 3' Untranslated Region of Enterovirus D68 To Inhibit Viral Replication.
Journal of virology
Cheng J,Gao S,Zhu C,Liu S,Li J,Kang J,Wang Z,Wang T
Tue Mar 17 00:00:00 EDT 2020
Typical Stress Granule Proteins Interact with the 3' Untranslated Region of Enterovirus D68 To Inhibit Viral Replication.
Journal of virology
Cheng J,Gao S,Zhu C,Liu S,Li J,Kang J,Wang Z,Wang T
Tue Mar 17 00:00:00 EDT 2020
MAFG-AS1 promotes tumor progression via regulation of the HuR/PTBP1 axis in bladder urothelial carcinoma.
Clinical and translational medicine
Xiao M,Liu J,Xiang L,Zhao K,He D,Zeng Q,Zhang Q,Xie D,Deng M,Zhu Y,Zhang Y,Liu Y,Bo H,Liu X,Chen X,Gong L,Bao Y,Hu Y,Cheng Y,Deng L,Zhu R,Xing X,Zhou M,Xiong W,Zhou Y,Zhou J,Li X,Cao K
Tue Dec 01 00:00:00 EST 2020
MAFG-AS1 promotes tumor progression via regulation of the HuR/PTBP1 axis in bladder urothelial carcinoma.
Clinical and translational medicine
Xiao M,Liu J,Xiang L,Zhao K,He D,Zeng Q,Zhang Q,Xie D,Deng M,Zhu Y,Zhang Y,Liu Y,Bo H,Liu X,Chen X,Gong L,Bao Y,Hu Y,Cheng Y,Deng L,Zhu R,Xing X,Zhou M,Xiong W,Zhou Y,Zhou J,Li X,Cao K
Tue Dec 01 00:00:00 EST 2020
MAFG-AS1 promotes tumor progression via regulation of the HuR/PTBP1 axis in bladder urothelial carcinoma.
Clinical and translational medicine
Xiao M,Liu J,Xiang L,Zhao K,He D,Zeng Q,Zhang Q,Xie D,Deng M,Zhu Y,Zhang Y,Liu Y,Bo H,Liu X,Chen X,Gong L,Bao Y,Hu Y,Cheng Y,Deng L,Zhu R,Xing X,Zhou M,Xiong W,Zhou Y,Zhou J,Li X,Cao K
Tue Dec 01 00:00:00 EST 2020
MAFG-AS1 promotes tumor progression via regulation of the HuR/PTBP1 axis in bladder urothelial carcinoma.
Clinical and translational medicine
Xiao M,Liu J,Xiang L,Zhao K,He D,Zeng Q,Zhang Q,Xie D,Deng M,Zhu Y,Zhang Y,Liu Y,Bo H,Liu X,Chen X,Gong L,Bao Y,Hu Y,Cheng Y,Deng L,Zhu R,Xing X,Zhou M,Xiong W,Zhou Y,Zhou J,Li X,Cao K
Tue Dec 01 00:00:00 EST 2020
LncRNA MALAT1 affects the migration and invasion of trophoblast cells by regulating FOS expression in early-onset preeclampsia.
Pregnancy hypertension
Li Q,Wang T,Huang S,Zuo Q,Jiang Z,Yang N,Sun L
Wed Jul 01 00:00:00 EDT 2020
Exosomal circSHKBP1 promotes gastric cancer progression via regulating the miR-582-3p/HUR/VEGF axis and suppressing HSP90 degradation.
Molecular cancer
Xie M,Yu T,Jing X,Ma L,Fan Y,Yang F,Ma P,Jiang H,Wu X,Shu Y,Xu T
Mon Jun 29 00:00:00 EDT 2020
Exosomal circSHKBP1 promotes gastric cancer progression via regulating the miR-582-3p/HUR/VEGF axis and suppressing HSP90 degradation.
Molecular cancer
Xie M,Yu T,Jing X,Ma L,Fan Y,Yang F,Ma P,Jiang H,Wu X,Shu Y,Xu T
Mon Jun 29 00:00:00 EDT 2020
Exosomal circSHKBP1 promotes gastric cancer progression via regulating the miR-582-3p/HUR/VEGF axis and suppressing HSP90 degradation.
Molecular cancer
Xie M,Yu T,Jing X,Ma L,Fan Y,Yang F,Ma P,Jiang H,Wu X,Shu Y,Xu T
Mon Jun 29 00:00:00 EDT 2020
Plasticity of nuclear and cytoplasmic stress responses of RNA-binding proteins.
Nucleic acids research
Backlund M,Stein F,Rettel M,Schwarzl T,Perez-Perri JI,Brosig A,Zhou Y,Neu-Yilik G,Hentze MW,Kulozik AE
Thu May 21 00:00:00 EDT 2020
Plasticity of nuclear and cytoplasmic stress responses of RNA-binding proteins.
Nucleic acids research
Backlund M,Stein F,Rettel M,Schwarzl T,Perez-Perri JI,Brosig A,Zhou Y,Neu-Yilik G,Hentze MW,Kulozik AE
Thu May 21 00:00:00 EDT 2020
An oncopeptide regulates m6A recognition by the m6A reader IGF2BP1 and tumorigenesis.
Nature communications
Zhu S,Wang JZ,Chen D,He YT,Meng N,Chen M,Lu RX,Chen XH,Zhang XL,Yan GR
Fri Apr 03 00:00:00 EDT 2020
Fast and unbiased purification of RNA-protein complexes after UV cross-linking.
Methods (San Diego, Calif.)
Urdaneta EC,Beckmann BM
Mon Jun 01 00:00:00 EDT 2020
Hepatic HuR modulates lipid homeostasis in response to high-fat diet.
Nature communications
Zhang Z,Zong C,Jiang M,Hu H,Cheng X,Ni J,Yi X,Jiang B,Tian F,Chang MW,Su W,Zhu L,Li J,Xiang X,Miao C,Gorospe M,de Cabo R,Dou Y,Ju Z,Yang J,Jiang C,Yang Z,Wang W
Tue Jun 16 00:00:00 EDT 2020
Hepatic HuR modulates lipid homeostasis in response to high-fat diet.
Nature communications
Zhang Z,Zong C,Jiang M,Hu H,Cheng X,Ni J,Yi X,Jiang B,Tian F,Chang MW,Su W,Zhu L,Li J,Xiang X,Miao C,Gorospe M,de Cabo R,Dou Y,Ju Z,Yang J,Jiang C,Yang Z,Wang W
Tue Jun 16 00:00:00 EDT 2020
Nuclear retention element recruits U1 snRNP components to restrain spliced lncRNAs in the nucleus.
RNA biology
Azam S,Hou S,Zhu B,Wang W,Hao T,Bu X,Khan M,Lei H
Thu Aug 01 00:00:00 EDT 2019
LncRNA FENDRR attenuates adriamycin resistance via suppressing MDR1 expression through sponging HuR and miR-184 in chronic myelogenous leukaemia cells.
FEBS letters
Zhang F,Ni H,Li X,Liu H,Xi T,Zheng L
Thu Aug 01 00:00:00 EDT 2019
Zika Virus Subverts Stress Granules To Promote and Restrict Viral Gene Expression.
Journal of virology
Bonenfant G,Williams N,Netzband R,Schwarz MC,Evans MJ,Pager CT
Sat Jun 15 00:00:00 EDT 2019
Zika Virus Subverts Stress Granules To Promote and Restrict Viral Gene Expression.
Journal of virology
Bonenfant G,Williams N,Netzband R,Schwarz MC,Evans MJ,Pager CT
Sat Jun 15 00:00:00 EDT 2019
Stress granules regulate stress-induced paraspeckle assembly.
The Journal of cell biology
An H,Tan JT,Shelkovnikova TA
Mon Dec 02 00:00:00 EST 2019
LncRNA-Safe contributes to cardiac fibrosis through Safe-Sfrp2-HuR complex in mouse myocardial infarction.
Theranostics
Hao K,Lei W,Wu H,Wu J,Yang Z,Yan S,Lu XA,Li J,Xia X,Han X,Deng W,Zhong G,Zhao ZA,Hu S
Fri Sep 11 00:00:00 EDT 2020
Conditional deletion of smooth muscle Cullin-3 causes severe progressive hypertension.
JCI insight
Agbor LN,Nair AR,Wu J,Lu KT,Davis DR,Keen HL,Quelle FW,McCormick JA,Singer JD,Sigmund CD
Tue Jun 11 00:00:00 EDT 2019
Purification of cross-linked RNA-protein complexes by phenol-toluol extraction.
Nature communications
Urdaneta EC,Vieira-Vieira CH,Hick T,Wessels HH,Figini D,Moschall R,Medenbach J,Ohler U,Granneman S,Selbach M,Beckmann BM
Fri Mar 01 00:00:00 EST 2019
Zc3h13 Regulates Nuclear RNA m6A Methylation and Mouse Embryonic Stem Cell Self-Renewal.
Molecular cell
Wen J,Lv R,Ma H,Shen H,He C,Wang J,Jiao F,Liu H,Yang P,Tan L,Lan F,Shi YG,He C,Shi Y,Diao J
Thu Mar 15 00:00:00 EDT 2018
Zc3h13 Regulates Nuclear RNA m6A Methylation and Mouse Embryonic Stem Cell Self-Renewal.
Molecular cell
Wen J,Lv R,Ma H,Shen H,He C,Wang J,Jiao F,Liu H,Yang P,Tan L,Lan F,Shi YG,He C,Shi Y,Diao J
Thu Mar 15 00:00:00 EDT 2018
Discovery of RNA-binding proteins and characterization of their dynamic responses by enhanced RNA interactome capture.
Nature communications
Perez-Perri JI,Rogell B,Schwarzl T,Stein F,Zhou Y,Rettel M,Brosig A,Hentze MW
Tue Oct 23 00:00:00 EDT 2018
Discovery of RNA-binding proteins and characterization of their dynamic responses by enhanced RNA interactome capture.
Nature communications
Perez-Perri JI,Rogell B,Schwarzl T,Stein F,Zhou Y,Rettel M,Brosig A,Hentze MW
Tue Oct 23 00:00:00 EDT 2018
Dysregulation of tristetraprolin and human antigen R promotes gastric cancer progressions partly by upregulation of the high-mobility group box 1.
Scientific reports
Wang H,Chen Y,Guo J,Shan T,Deng K,Chen J,Cai L,Zhou H,Zhao Q,Jin S,Xia J
Fri May 04 00:00:00 EDT 2018
Dysregulation of tristetraprolin and human antigen R promotes gastric cancer progressions partly by upregulation of the high-mobility group box 1.
Scientific reports
Wang H,Chen Y,Guo J,Shan T,Deng K,Chen J,Cai L,Zhou H,Zhao Q,Jin S,Xia J
Fri May 04 00:00:00 EDT 2018
Dcp1a and GW182 Induce Distinct Cellular Aggregates and Have Different Effects on microRNA Pathway.
DNA and cell biology
Wang X,Chang L,Wang H,Su A,Wu Z
Sat Jul 01 00:00:00 EDT 2017
Dcp1a and GW182 Induce Distinct Cellular Aggregates and Have Different Effects on microRNA Pathway.
DNA and cell biology
Wang X,Chang L,Wang H,Su A,Wu Z
Sat Jul 01 00:00:00 EDT 2017
E2F-1 targets miR-519d to regulate the expression of the ras homolog gene family member C.
Oncotarget
Sang XB,Zong ZH,Wang LL,Wu DD,Chen S,Liu BL,Zhao Y
Tue Feb 28 00:00:00 EST 2017
E2F-1 targets miR-519d to regulate the expression of the ras homolog gene family member C.
Oncotarget
Sang XB,Zong ZH,Wang LL,Wu DD,Chen S,Liu BL,Zhao Y
Tue Feb 28 00:00:00 EST 2017
Inhibition of COX2 enhances the chemosensitivity of dichloroacetate in cervical cancer cells.
Oncotarget
Li B,Li X,Xiong H,Zhou P,Ni Z,Yang T,Zhang Y,Zeng Y,He J,Yang F,Zhang N,Wang Y,Zheng Y,He F
Tue Aug 01 00:00:00 EDT 2017
Inhibition of COX2 enhances the chemosensitivity of dichloroacetate in cervical cancer cells.
Oncotarget
Li B,Li X,Xiong H,Zhou P,Ni Z,Yang T,Zhang Y,Zeng Y,He J,Yang F,Zhang N,Wang Y,Zheng Y,He F
Tue Aug 01 00:00:00 EDT 2017
A long non-coding RNA, APOA4-AS, regulates APOA4 expression depending on HuR in mice.
Nucleic acids research
Qin W,Li X,Xie L,Li S,Liu J,Jia L,Dong X,Ren X,Xiao J,Yang C,Zhou Y,Chen Z
Wed Jul 27 00:00:00 EDT 2016
A long non-coding RNA, APOA4-AS, regulates APOA4 expression depending on HuR in mice.
Nucleic acids research
Qin W,Li X,Xie L,Li S,Liu J,Jia L,Dong X,Ren X,Xiao J,Yang C,Zhou Y,Chen Z
Wed Jul 27 00:00:00 EDT 2016
Long Noncoding RNA MALAT1 Promotes Aggressive Pancreatic Cancer Proliferation and Metastasis via the Stimulation of Autophagy.
Molecular cancer therapeutics
Li L,Chen H,Gao Y,Wang YW,Zhang GQ,Pan SH,Ji L,Kong R,Wang G,Jia YH,Bai XW,Sun B
Thu Sep 01 00:00:00 EDT 2016
Long Noncoding RNA MALAT1 Promotes Aggressive Pancreatic Cancer Proliferation and Metastasis via the Stimulation of Autophagy.
Molecular cancer therapeutics
Li L,Chen H,Gao Y,Wang YW,Zhang GQ,Pan SH,Ji L,Kong R,Wang G,Jia YH,Bai XW,Sun B
Thu Sep 01 00:00:00 EDT 2016
Long Noncoding RNA MALAT1 Promotes Aggressive Pancreatic Cancer Proliferation and Metastasis via the Stimulation of Autophagy.
Molecular cancer therapeutics
Li L,Chen H,Gao Y,Wang YW,Zhang GQ,Pan SH,Ji L,Kong R,Wang G,Jia YH,Bai XW,Sun B
Thu Sep 01 00:00:00 EDT 2016
Long Noncoding RNA MALAT1 Promotes Aggressive Pancreatic Cancer Proliferation and Metastasis via the Stimulation of Autophagy.
Molecular cancer therapeutics
Li L,Chen H,Gao Y,Wang YW,Zhang GQ,Pan SH,Ji L,Kong R,Wang G,Jia YH,Bai XW,Sun B
Thu Sep 01 00:00:00 EDT 2016
C1QBP negatively regulates the activation of oncoprotein YBX1 in the renal cell carcinoma as revealed by interactomics analysis.
Journal of proteome research
Wang Y,Yue D,Xiao M,Qi C,Chen Y,Sun D,Zhang N,Chen R
Fri Feb 06 00:00:00 EST 2015
C1QBP negatively regulates the activation of oncoprotein YBX1 in the renal cell carcinoma as revealed by interactomics analysis.
Journal of proteome research
Wang Y,Yue D,Xiao M,Qi C,Chen Y,Sun D,Zhang N,Chen R
Fri Feb 06 00:00:00 EST 2015
HuR antagonizes the effect of an intronic pyrimidine-rich sequence in regulating WT1 +/-KTS isoforms.
RNA biology
Li H,Hou S,Hao T,Azam S,Liu C,Shi L,Lei H
Wed Oct 12 00:00:00 EDT 2016
HuR antagonizes the effect of an intronic pyrimidine-rich sequence in regulating WT1 +/-KTS isoforms.
RNA biology
Li H,Hou S,Hao T,Azam S,Liu C,Shi L,Lei H
Wed Oct 12 00:00:00 EDT 2016
H19 lncRNA alters DNA methylation genome wide by regulating S-adenosylhomocysteine hydrolase.
Nature communications
Zhou J,Yang L,Zhong T,Mueller M,Men Y,Zhang N,Xie J,Giang K,Chung H,Sun X,Lu L,Carmichael GG,Taylor HS,Huang Y
Mon Dec 21 00:00:00 EST 2015
H19 lncRNA alters DNA methylation genome wide by regulating S-adenosylhomocysteine hydrolase.
Nature communications
Zhou J,Yang L,Zhong T,Mueller M,Men Y,Zhang N,Xie J,Giang K,Chung H,Sun X,Lu L,Carmichael GG,Taylor HS,Huang Y
Mon Dec 21 00:00:00 EST 2015
The Structure of the Cyprinid herpesvirus 3 ORF112-Zα·Z-DNA Complex Reveals a Mechanism of Nucleic Acids Recognition Conserved with E3L, a Poxvirus Inhibitor of Interferon Response.
The Journal of biological chemistry
Kuś K,Rakus K,Boutier M,Tsigkri T,Gabriel L,Vanderplasschen A,Athanasiadis A
Fri Dec 25 00:00:00 EST 2015
The Structure of the Cyprinid herpesvirus 3 ORF112-Zα·Z-DNA Complex Reveals a Mechanism of Nucleic Acids Recognition Conserved with E3L, a Poxvirus Inhibitor of Interferon Response.
The Journal of biological chemistry
Kuś K,Rakus K,Boutier M,Tsigkri T,Gabriel L,Vanderplasschen A,Athanasiadis A
Fri Dec 25 00:00:00 EST 2015
AUF1 is recruited to the stress granules induced by coxsackievirus B3.
Virus research
Wu S,Lin L,Zhao W,Li X,Wang Y,Si X,Wang T,Wu H,Zhai X,Zhong X,Gao S,Tong L,Xu Z,Zhong Z
Tue Nov 04 00:00:00 EST 2014
AUF1 is recruited to the stress granules induced by coxsackievirus B3.
Virus research
Wu S,Lin L,Zhao W,Li X,Wang Y,Si X,Wang T,Wu H,Zhai X,Zhong X,Gao S,Tong L,Xu Z,Zhong Z
Tue Nov 04 00:00:00 EST 2014
HuR and TIA1/TIAL1 are involved in regulation of alternative splicing of SIRT1 pre-mRNA.
International journal of molecular sciences
Zhao W,Zhao J,Hou M,Wang Y,Zhang Y,Zhao X,Zhang C,Guo D
Thu Feb 20 00:00:00 EST 2014
HuR and TIA1/TIAL1 are involved in regulation of alternative splicing of SIRT1 pre-mRNA.
International journal of molecular sciences
Zhao W,Zhao J,Hou M,Wang Y,Zhang Y,Zhao X,Zhang C,Guo D
Thu Feb 20 00:00:00 EST 2014
Post-transcriptional up-regulation of PDGF-C by HuR in advanced and stressed breast cancer.
International journal of molecular sciences
Luo NA,Qu YQ,Yang GD,Wang T,Li RL,Jia LT,Dong R
Thu Nov 06 00:00:00 EST 2014
Post-transcriptional up-regulation of PDGF-C by HuR in advanced and stressed breast cancer.
International journal of molecular sciences
Luo NA,Qu YQ,Yang GD,Wang T,Li RL,Jia LT,Dong R
Thu Nov 06 00:00:00 EST 2014
Post-transcriptional up-regulation of PDGF-C by HuR in advanced and stressed breast cancer.
International journal of molecular sciences
Luo NA,Qu YQ,Yang GD,Wang T,Li RL,Jia LT,Dong R
Thu Nov 06 00:00:00 EST 2014
Post-transcriptional up-regulation of PDGF-C by HuR in advanced and stressed breast cancer.
International journal of molecular sciences
Luo NA,Qu YQ,Yang GD,Wang T,Li RL,Jia LT,Dong R
Thu Nov 06 00:00:00 EST 2014
Protease 2A induces stress granule formation during coxsackievirus B3 and enterovirus 71 infections.
Virology journal
Wu S,Wang Y,Lin L,Si X,Wang T,Zhong X,Tong L,Luan Y,Chen Y,Li X,Zhang F,Zhao W,Zhong Z
Thu Nov 20 00:00:00 EST 2014
Protease 2A induces stress granule formation during coxsackievirus B3 and enterovirus 71 infections.
Virology journal
Wu S,Wang Y,Lin L,Si X,Wang T,Zhong X,Tong L,Luan Y,Chen Y,Li X,Zhang F,Zhao W,Zhong Z
Thu Nov 20 00:00:00 EST 2014