Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSNGYEDHM AEDCRGDIGR TNLIVNYLPQ NMTQDELRSL FSSIGEVESA KLIRDKVAGH SLGYGFVNYV TAKDAERAIN TLNGLRLQSK TIKVSYARPS SEVIKDANLY ISGLPRTMTQ KDVEDMFSRF GRIINSRVLV DQTTGLSRGV AFIRFDKRSE AEEAITSFNG HKPPGSSEPI TVKFAANPNQ NKNVALLSQL YHSPARRFGG PVHHQAQRFR FSPMGVDHMS GLSGVNVPGN ASSGWCIFIY NLGQDADEGI LWQMFGPFGA VTNVKVIRDF NTNKCKGFGF VTMTNYEEAA MAIASLNGYR LGDKILQVSF KTNKSHK (1-326 aa encoded by BC003376)
The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; Elav-like generic protein; ELAV-like protein 1; embryonic lethal, abnormal vision, drosophila, homolog-like 1; Hu antigen R; HU-antigen A; Hu-antigen R; Human antigen R; HuR; MelG
Gene Aliases: 2410055N02Rik; ELAV1; ELAVL1; Elra; Hua; HUR; MelG; W91709
UniProt ID: (Human) Q15717, (Mouse) P70372, (Rat) B5DF91
Entrez Gene ID: (Human) 1994, (Mouse) 15568, (Rat) 363854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support