Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGTPKPRIL PWLVSQLDLG QLEGVAWVNK SRTRFRIPWK HGLRQDAQQE DFGIFQAWAE ATGAYVPGRD KPDLPTWKRN FRSALNRKEG LRLAEDRSKD PHDPHKIYEF VNSGVGDFSQ PDTSPDTNGG GSTSDTQEDI LDELLGNMVL APLPDPGPPS LAVAPEPCPQ PLRSPSLDNP TPFPNLGPSE NPLKRLLVPG EEWEFEVTAF YRGRQVFQQT ISCPEGLRLV GSEVGDRTLP GWPVTLPDPG MSLTDRGVMS YVRHVLSCLG GGLALWRAGQ WLWAQRLGHC HTYWAVSEEL LPNSGHGPDG EVPKDKEGGV FDLGPFIVGS WAPRSDYLHG RKRTLTTLCP LVLCGGVMAP GPAVDQEARD GQGCAHVPQG LGRNGPGRGC LLPGEYCGPA HFQQPPTLPH LRPVQGLPAG LGGGHGFPGP WGDLSPRSSW CASNPPVPHH LNQ (1-452 aa encoded by BC009395)
IFR3 (interferon regulatory factor 3) is a key transcriptional regulator of type I interferon (IFN)-dependent immune responses. It plays a critical role in the innate immune response against DNA and RNA viruses. IFR3 regulates the transcription of type I IFN genes (IFN-alpha and IFN-beta) and IFN-stimulated genes (ISG) by binding to an interferon-stimulated response element (ISRE) in their promoters. It acts as a more potent activator of the IFN-beta gene than the IFN-alpha gene and plays a critical role in both the early and late phases of the IFNA/B gene induction. IFR3 is found in an inactive form in the cytoplasm of uninfected cells and following viral infection, double-stranded RNA, or toll-like receptor signaling. It is phosphorylated by IKBKE and TBK1 kinases. This induces a conformational change, leading to its dimerization and nuclear localization and association with CREB binding protein to form dsRNA-activated factor 1 (DRAF1), a complex which activates the transcription of the type I IFN and ISG genes. Mutations in the gene can result in encephalopathy, acute, infection-induced, herpes-specific 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interferon regulatory factor 3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3
Gene Aliases: IIAE7; IRF3
UniProt ID: (Human) Q14653
Entrez Gene ID: (Human) 3661
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support