Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IVMNDQVLEP QNVDPSMVQM TFLDDVVHSL LKGENIGITS RRRSRANQNV NAVHSHYTRA QANSPRPAMN SQAAVPKQNT HQQQQQRSIR PNKRKGSD
The jumonji domain containing 1C protein (JMJD1C) was initially discovered in silico, and later suggested to be a candidate gene for autism. Like the related proteins JMJD1A and JMJD1B, JMJD1C is a histone H3K9 demethylase implicated in the nuclear hormone receptor-based transcriptional regulation. JMJD1C mRNA is highly expressed in undifferentiated embryonic stem (ES) cells as well as pancreatic islet, diffuse-type gastric cancer, and other tissues and tumors. The JMJD1C gene promoter contain bHLH-, AP-1-, and POU5F1-binding sites, and as preferential expression of POU5F1 has been reported in ES cells, pancreatic islet, and diffuse-type gastric cancer, it has been suggested that POU5F1-mediated expression of JMJD1C reactivates previously silenced genes in ES cells and diffuse-type gastric cancer. At least three isoforms of JMJD1C are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761F0118; FLJ14374; Jumonji domain-containing protein 1C; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2; thyroid hormone receptor interactor 8; Thyroid receptor-interacting protein 8; TR-interacting protein 8
Gene Aliases: JHDM2C; JMJD1C; KIAA1380; TRIP-8; TRIP8
UniProt ID: (Human) Q15652
Entrez Gene ID: (Human) 221037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support