Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDSKCDSQ FYSVQVADST FTVLKRYQQL KPIGSGAQGI VCAAFDTVLG INVAVKKLSR PFQNQTHAKR AYRELVLLKC VNHKNIISLL NVFTPQKTLE EFQDVYLVME LMDANLCQVI HMELDHERMS YLLYQMLCGI KHLHSAGIIH RDLKPSNIVV KSDCTLKILD FGLARTACTN FMMTPYVVTR YYRAPEVILG MGYKENVDIW SVGCIMGELV KGCVIFQGTD HIDQWNKVIE QLGTPSAEFM KKLQPTVRNY VENRPKYPGI KFEELFPDWI FPSESERDKI KTSQARDLLS KMLVIDPDKR ISVDEALRHP YITVWYDPAE AEAPPPQIYD AQLEEREHAI EEWKELIYKE VMDWEERSKN GVVKDQPSDA AVSSNATPSQ SSSINDISSM STEQTLASDT DSSLDASTGP LEGCR (1-424 aa encoded by BC032539)
MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun kinase 2; c-Jun N-terminal kinase 2; JNK; JNK-55; JNK/SAPK alpha; JNK1; JNK1A2; JNK21B1/2; Jun kinase; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; mitogen activated protein kinase 9; Mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54-alpha; PRKM8; protein kinase, mitogen-activated 9; SAPK-alpha; SAPK1; SAPK1a; stress activated protein kinase alpha II; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: AI851083; JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984, (Rat) P49186, (Mouse) Q9WTU6
Entrez Gene ID: (Human) 5601, (Rat) 50658, (Mouse) 26420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support