Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCTKMEQPF YHDDSYTATG YGRAPGGLSL HDYKLLKPSL AVNLADPYRS LKAPGARGPG PEGGGGGSYF SGQGSDTGAS LKLASSELER LIVPNSNGVI TTTPTPPGQY FYPRGGGSGG GAGGAGGGVT EEQEGFADGF VKALDDLHKM NHVTPPNVSL GATGGPPAGP GGVYAGPEPP PVYTNLSSYS PASASSGGAG A (1-200 aa encoded by BC004250)
JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activator protein 1; Jun B; jun B proto-oncogene; Jun-B oncogene; MyD21; Oncogene Jun-B; Transcription factor AP-1 subunit JunB; transcription factor jun-B; Transcription factor JunB
Gene Aliases: AP-1; Jun-b; JUNB
UniProt ID: (Human) P17275, (Rat) P24898, (Mouse) P09450
Entrez Gene ID: (Human) 3726, (Rat) 24517, (Mouse) 16477
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support