Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MQSILYHGLI RDQACRRQTD YWQFVKDIRW LSPHSALHVE KFISVHENDQ SSADGASERA VAELWLQHSL QYHCLSAQLR PLLGDRQYIR KFYTDAAFLL SDAHVTAMLQ CLEAVEQNNP RLLAQIDASM FARKHESPLL VTKSQSLTAL PSSTYTPPNS YAQHSYFGSF SSLHQSVPNN GSERRSTSFP LSGPPRKPQE SRGHVSPAED QTIQAPPVSV SALARDSPLT PNEMSSSTLT SPIEASWVSS QNDSPGDASE GPEYLAIGNL DPRGRTASCQ SHSSNAESSS SNLFSSSSSQ KPDSAASSLG DQEGGGESQL SSVLRRSSFS EGQTLTVTSG AKKSHIRSHS DTSIASRGAP GNEEHRLLVS KMTLN
Two Beclin-1-interacting proteins, the run domain Beclin-1 interacting and cysteine-rich containing protein (Rubicon) and ATG14L, reciprocally regulate autophagy at different stages. Knockdown of Rubicon caused enhancement of autophagy while that of ATG14L caused a defect in autophagosome formation. Rubicon functions as part of a Beclin-1-PIK3C3-containing autophagy complex and is also an essential, positive regulator of the NAPDH oxidase complex. Upon microbial infection or TLR2 activation, Rubicon interacts with the CYBA subunit of the NAPDH oxidase complex, leading to a burst of reactive oxygen species and inflammatory cytokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Baron; Beclin-1 associated RUN domain containing protein; Rubicon; RUN domain and cysteine-rich domain containing, Beclin 1-interacting protein; run domain Beclin-1 interacting and cystein-rich containing protein; run domain Beclin-1 interacting and cysteine-rich containing protein; Run domain Beclin-1-interacting and cysteine-rich domain-containing protein; RUN domain protein as Beclin 1-interacting and cysteine-rich containing; rundataxin
Gene Aliases: KIAA0226; RUBCN; RUBICON; SCAR15
UniProt ID: (Human) Q92622
Entrez Gene ID: (Human) 9711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support