Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: METPEVPVG SLIDFGPEAP TSSPLEAPPP VLQDGDGSLG DGASESETTE SADSENDMGE SPSHPSWDQD RRSSSNESFS SNQSTESTQD EETLALRDFM RGYVEKIFSG GEDLDQEEKA KFGEYCSSEN GKGREWFARY VSAQRCNSKC VSEATFYRLV QSFAVVLFEC HQMDDFGPAK NLMTMCFTYY HIGKPQLLPP ESREKPAGSI DSYLKSANSW LAEKKDIAER LLKNTSARTE NVKGFFGGLE TKLKGPLARR NEEDENKPQE KRPRAVTAYS PEDEKKGEKI YLYTHLKQQP IW (1-301 aa encoded by BC030280)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein KIAA0513; uncharacterized protein KIAA0513 homolog
Gene Aliases: AW049007; KIAA0513; mKIAA0513
UniProt ID: (Mouse) Q8R0A7, (Human) O60268
Entrez Gene ID: (Mouse) 234797, (Human) 9764, (Rat) 307907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support