Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVCDNLNLPF RDEGFDAIIS IGVIHHFSTK QRRIRAIKEM ARVLVPGGQL MIYVWAMEQK NRRFEKQDVL VPWNRALCSQ LFSESSQSGR KRQCGYPERG HPYHPPCSEC SCSVCFKEQG GSKRSHSVGY EPAMARTCFA NISKEGEEEY GFYSTLGKSF RSWFFSRSLD ESTLRKQIER VRPLKNTEVW ASSTVTVQPS RHSSLDFDHQ EPFSTKEQSL DEEVFVESSS GKHLEWLRAP GTLKHLNGDH QGEMRRNGGG NFLDSTNTGV NCVDAGNIED DNPSASKILR RISAVDSTDF NPDDTMSVED PQTDVLDSTA FMRYYHVFRE GELCSLLKEN VSELRILSSG NDHGNWCIIA EKKGGCD
C8orf79 (chromosome 8 open reading frame 79) is a 454 amino acid protein that belongs to the methyltransferase superfamily and is encoded by a gene that maps to human chromosome 8p22. Consisting of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and are typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Probable tRNA methyltransferase 9-like protein; Probable tRNA methyltransferase 9B; RM9L
Gene Aliases: C8orf79; KIAA1456; TRM9L; TRMT9B
UniProt ID: (Human) Q9P272
Entrez Gene ID: (Human) 57604
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support