Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQRMIQQFA AEYTSKNSST QDPSQPNSTK NQSLPKASPV TTSPTAATTQ NPVLSKLLMA DQDSPLDLTV RKSQSEPSEQ DGVLDLSTKK SPCAGSTSLS HSPGCSSTQG NGRPGRPSQY RPDGLRSGDG VPPRSLQDGT REGFGHSTSL KVPLARSLQI SEELLSRNQL STAASLGPSG LQNHGQHLIL SREASWAKPH YEFNLSRMKF RGNGALSNIS DLPFLAENSA FPKMALQAKQ DGKKDVSHSS PVDLKIPQVR GMDLSWESRT GDQYSYSSLV MGSQTESALS KKLRAILPKQ SRKSMLDAGP DSWGSDAEQS TSGQPYPTSD QEGDPGSKQP RKKRGRYRQY NSEILEEAIS VVMSGKMSVS KAQSIYGIPH STLEYKVKER LGTLKNPPKK KMKLMSG (1-406 aa encoded by BC053359)
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LCoR; ligand dependent nuclear receptor corepressor; Ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; transcription factor MLR2
Gene Aliases: 3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; LCOR; mKIAA1795; MLR2; RGD1311704
UniProt ID: (Human) Q96JN0, (Mouse) Q6ZPI3
Entrez Gene ID: (Human) 84458, (Rat) 365462, (Mouse) 212391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support