Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LINGO4 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine rich repeat neuronal 6D; Leucine-rich repeat and immunoglobulin-like domain containing-NOGO receptor-interacting protein 4; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 4; Leucine-rich repeat neuronal protein 6D
Gene Aliases: A530050P17Rik; DAAT9248; LERN4; LINGO4; LRRN6D; PRO34002; RGD1562025; UNQ9248/PRO34002
UniProt ID: (Human) Q6UY18, (Mouse) Q149C3
Entrez Gene ID: (Human) 339398, (Rat) 499668, (Mouse) 320747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support