Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at 4° C prior to restoration. Restore with 1.0 mL of deionized water (or equivalent). For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Typically less than 1% cross-reactivity against other extracellular matrix proteins was detected by ELISA against purified standards. This antibody reacts with most mammalian Laminins and has negligible cross-reactivity with Type I, II, III, IV, V or VI Collagens or Fibronectin. Non-specific cross-reaction of anti-Laminin antibodies with other human serum proteins or non-Laminin extracellular matrix proteins is negligible.
Laminin antibodies are widely used to label blood vessels and basement membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Laminin A chain; Laminin B1 chain; Laminin B2 chain; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1 (formerly LAMB2); Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma
Gene Aliases: CLM; LAMA; LAMA1; LAMA2; LAMB1; LAMB2; LAMC1; LAMM; LIS5; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047
Entrez Gene ID: (Human) 284217, (Human) 3908, (Human) 3912, (Human) 3915
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support