Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in many other species.
Specificity of this antibody: Lingo1.
Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors (PubMed:14966521, PubMed:15694321). Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination (PubMed:15895088). Acts in conjunction with RTN4 and RTN4R in regulating neuronal precursor cell motility during cortical development. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine rich repeat neuronal 6A; leucine-rich repeat; Leucine-rich repeat and immunoglobulin domain-containing protein 1; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1; leucine-rich repeat neuronal 6A; Leucine-rich repeat neuronal protein 1; Leucine-rich repeat neuronal protein 6A
Gene Aliases: 4930471K13Rik; AV148400; LERN1; LINGO-1; LINGO1; LRRN6A; UNQ201; UNQ201/PRO227
UniProt ID: (Human) Q96FE5, (Mouse) Q9D1T0
Entrez Gene ID: (Human) 84894, (Rat) 315691, (Mouse) 235402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support