Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LY6A is included in a family of proteins that are highly homologous, low molecular weight glycophosphotidylinositol-linked proteins expressed on hematopoietic cells and lymphoid cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ly-6A.2/Ly-6E.1; Lymphocyte antigen 6A-2/6E-1; SCA-1; Stem cell antigen 1; T-cell-activating protein; TAP
Gene Aliases: Ly-6A.2; Ly-6A/E; Ly-6E.1; Ly6; Ly6a; Sca-1; Sca1; TAP
UniProt ID: (Mouse) P05533
Entrez Gene ID: (Mouse) 110454
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support