Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KARSAPKPS QASGHFSVEL VRGYAGFGLT LGGGRDVAGD TPLAVRGLLK DGPAQRCGRL EVGDLVLHIN GESTQGLTHA QAVERIRAGG PQLHLVIRRP LETHPGKPRG VGEPRKGVDR SPDPGGPEVT GSRSSSTSLV QHPPSRTTLK KTRGSPEPSP EAAADGPTVS PPERRAEDPN DQIPGSPGPW LVPSEERLSR (43-241 aa encoded by BC114943)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PDZ domain containing X chromosome; PDZ domain containing, X chromosome; PDZ domain-containing protein MAGIX
Gene Aliases: JM10; MAGIX; PDZX; Sfc18
UniProt ID: (Human) Q9H6Y5, (Mouse) Q4KL35
Entrez Gene ID: (Human) 79917, (Mouse) 54634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support