Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
MAGT1 encodes a magnesium cation transporter protein that localizes to the cell membrane. This protein also associates with N-oligosaccharyl transferase and therefore may have a role in N-glycosylation. Mutations in this gene cause mental retardation X-linked type 95 (MRX95). This gene may have multiple in-frame translation initiation sites, one of which would encode a shorter protein with an N-terminus containing a signal peptide at amino acids 1-29.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit MAGT1; IAP; Implantation-associated protein; Magnesium transporter protein 1; MagT1; Oligosaccharyl transferase subunit MAGT1; oligosaccharyltransferase 3 homolog B
Gene Aliases: 2410001C15Rik; 2610529C04Rik; 2810482I07Rik; bA217H1.1; IAG2; IAP; MAGT1; MRX95; OST3B; PRO0756; PSEC0084; UNQ628/PRO1244; XMEN
UniProt ID: (Human) D3DTE3, (Rat) O35777, (Mouse) Q9CQY5
Entrez Gene ID: (Human) 84061, (Rat) 116967, (Mouse) 67075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support