Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Sequence of this protein is as follows: MSTASAASSS SSSSAGEMIE APSQVLNFEE IDYKEIEVEE VVGRGAFGVV CKAKWRAKDV AIKQIESESE RKAFIVELRQ LSRVNHPNIV KLYGACLNPV CLVMEYAEGG SLYNVLHGAE PLPYYTAAHA MSWCLQCSQG VAYLHSMQPK ALIHRDLKPP NLLLVAGGTV LKICDFGTAC DIQTHMTNNK GSAAWMAPEV FEGSNYSEKC DVFSWGIILW EVITRRKPFD EIGGPAFRIM WAVHNGTRPP LIKNLPKPIE SLMTRCWSKD PSQRPSMEEI VKIMTHLMRY FPGADEPLQY PCQYSDEGQS NSATSTGSFM DIASTNTSNK SDTNMEQVPA TNDTIKRLES KLLKNQAKQQ SESGRLSLGA SRGSSVESLP PTSEGKRMSA DMSEIEARIA ATTGNGQPRR RSIQDLTVTG TEPGQVSSRS SSPSVRMITT SGPTSEKPTR SHPWTPDDST DTNGSDNSIP MAYLTLDHQL QPLAPCPNSK ESMAVFEQHC KMAQEYMKVQ TEIALLLQRK QELVAELDQD EKDQQNTSRL VQEHKKLLDE NKSLSTYYQQ CKKQLEVIRS QQQKRQGTS
TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M3K7; Mitogen-activated protein kinase kinase kinase 7; RP1-154G14.1; TGF-beta activated kinase 1; TGF-beta-activated kinase 1; Transforming growth factor-beta-activated kinase 1
Gene Aliases: MAP3K7; MEKK7; TAK1; TGF1a
UniProt ID: (Human) O43318
Entrez Gene ID: (Human) 6885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support