Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80K protein; 80K-L protein; FLJ14368; FLJ90045; MARCKS; MARCS; Myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; protein kinase C substrate 80 kDa protein; Protein kinase C substrate, 80 kDa protein, light chain
Gene Aliases: 80K-L; KINC; MACS; MARCKS; PKCSL; PRKCSL
UniProt ID: (Human) P29966, (Mouse) P26645
Entrez Gene ID: (Human) 4082, (Rat) 25603, (Mouse) 17118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support