Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QAAGPAIPTS NSYSKKTQSN NAENKRPEED RESGRKASST AKVPASPLPG LERKKTTPTP STNSVLSTST NRSRNSPLLE RASLGQASIQ NGKDSLTMPG
MARK2 is a serine/threonine protein kinase involved in the control of cell polarity and microtubule stability. Several cDNA clones have been isolated that encoded two isoforms of the human ser/thr protein kinase EMK1. These isoforms were characterized by the presence of a 162-bp alternative exon that gave rise to two forms, one containing the exon and the other one lacking it. Both forms were found to be coexpressed in a number of selected cell lines and tissue samples. The human EMK1 was shown to be encoded by a single mRNA ubiquitously expressed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELKL motif kinase; ELKL motif kinase 1; EMK-1; EMK1; MAP/microtubule affinity-regulating kinase 2; MARK2; mark2 {ECO:0000312|RGD:708483}; MGC99619; OTTHUMP00000218338; OTTHUMP00000218339; OTTHUMP00000218340; OTTHUMP00000218343; Par-1b; PAR1 homolog; PAR1 homolog b; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; Serine/threonine-protein kinase MARK2; testicular tissue protein Li 117
Gene Aliases: Emk; EMK-1; EMK1; MARK2; PAR-1; Par-1b; Par1b
UniProt ID: (Human) Q7KZI7, (Rat) O08679, (Mouse) Q05512
Entrez Gene ID: (Human) 2011, (Rat) 60328, (Mouse) 13728
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support