Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mediates the octanoylation of ghrelin at 'Ser-3'. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ghrelin O-acyltransferase; ghrelin O-acyltransferase {ECO:0000303|PubMed:18443287}; mboat4 {ECO:0000312|EMBL:ACB05875.1}; Membrane-bound O-acyltransferase domain-containing protein 4; membrane-bound O-acyltransferase domain-containing protein 4 {ECO:0000312|RGD:1566335}; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4
Gene Aliases: FKSG89; Gm171; GOAT; MBOAT4; OACT4; RGD1566335
UniProt ID: (Human) Q96T53, (Rat) B1Q005, (Mouse) P0C7A3
Entrez Gene ID: (Human) 619373, (Rat) 306515, (Mouse) 234155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support