Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWAVLRLALR PCARASPAGP RAYHGDSVAS LGTQPDLGSA LYQENYKQMK ALVNQLHERV EHIKLGGGEK ARALHISRGK LLPRERIDNL IDPGSPFLEL SQFAGYQLYD NEEVPGGGII TGIGRVSGVE CMIIANDATV KGGAYYPVTV KKQLRAQEIA MQNRLPCIYL VDSGGAYLPR QADVFPDRDH FGRTFYNQAI MSSKNIAQIA VVMGSCTAGG AYVPAMADEN IIVRKQGTIF LAGPPLVKAA TGEEVSAEDL GGADLHCRKS GVSDHWALDD HHALHLTRKV VRNLNYQKKL DVTIEPSEEP LFPADELYGI VGANLKRSFD VREVIARIVD GSRFTEFKAF YGDTLVTGFA RIFGYPVGIV GNNGVLFSES AKKGTHFVQL CCQRNIPLLF LQNITGFMVG REYEAEGIAK DGAKMVAAVA CAQVPKITLI IGGSYGAGNY GMCGRAYSPR FLYIWPNARI SVMGGEQAAN VLATITKDQR AREGKQFSSA DEAALKEPII KKFEEEGNPY YSSARVWDDG IIDPADTRLV LGLSFSAALN APIEKTDFGI FRM
COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta; biotin carboxylase; MCCase subunit beta; meth; methylcrotonoyl-CoA carboxylase 2 (beta); Methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 2 (beta); non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; testicular secretory protein Li 29
Gene Aliases: 4930552N12Rik; MCCB; MCCC2
UniProt ID: (Human) Q9HCC0, (Mouse) Q3ULD5
Entrez Gene ID: (Human) 64087, (Mouse) 78038
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support