Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Application for antibody includes Stable Isotope Standards and Capture by Anti-Peptide Antibodies (SISCAPA).
Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: corneal keratan sulfate proteoglycan; Mimecan; mimecan proteoglycan; OIF; Osteoglycin; Osteoinductive factor
Gene Aliases: 3110079A16Rik; mimecan; mimican; OG; OGN; OIF; SLRR3A
UniProt ID: (Human) P20774, (Mouse) Q62000
Entrez Gene ID: (Human) 4969, (Rat) 291015, (Mouse) 18295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support