Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLLLEDIKKV ITPSKKNYND CIQGALLHQD VNTAWNLYQE LLGHDIVPML ETLKAFFDFG KDIKDDNYSN KLLDILSYLR NNQLYPGESF AHSIKTWFES VPGKQWKGQF TTVRKSGQCS GCGKTIESIQ LSPEEYECLK GKIMRDVIDG GDQYRKTTPQ ELKRFENFIK SRPPFDVVID GLNVAKMFPK VRESQLLLNV VSQLAKRNLR LLVLGRKHML RRSSQWSRDE MEEVQKQASC FFADDISEDD PFLLYATLHS GNHCRFITRD LMRDHKACLP DAKTQRLFFK WQQGHQLAIV NRFPGSKLTF QRILSYDTVV QTTGDSWHIP YDEDLVERCS CEVPTKWLCL HQKT
KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5'-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3. 5% of human cellular DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial ribonuclease P catalytic subunit; Mitochondrial ribonuclease P protein 3; Mitochondrial RNase P protein 3; mitochondrial RNase P subunit 3; Protein only RNase P catalytic subunit; proteinaceous RNase P
Gene Aliases: KIAA0391; mKIAA0391; MRPP3; PRORP
UniProt ID: (Mouse) Q8JZY4, (Human) O15091
Entrez Gene ID: (Mouse) 66132, (Human) 9692
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support