Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Rat large intestine.
Immunogen sequence: IFSTQNMNLN GENLENSGST LSLNPLPSHS VLNESKCFIK DNTLALESNY PEDSLLSSSL NVASDSIAGC SSLNQKQKEL LESECVEAQF SEAPVDLDAS EPQACLNLPG LDLPGTGGDQ KSTRVSDVFL P
Has 5'-polynucleotide kinase and nicking endonuclease activity. May play a role in DNA repair or recombination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCL-3-binding protein; N4BP2; NEDD4 binding protein 2; NEDD4-binding protein 2
Gene Aliases: B3BP; KIAA1413; N4BP2; RGD1565738
UniProt ID: (Human) Q86UW6
Entrez Gene ID: (Human) 55728, (Rat) 305342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support