Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Functions as a transcriptional repressor through its association with the NuRD complex. Recruits the NuRD complex to the promoter of MDM2, leading to the repression of MDM2 transcription and subsequent stability of p53/TP53. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain containing 9; BTB (POZ) domain containing 14A; BTB/POZ domain-containing protein 14A; MGC23427; NAC-2; NACC family member 2, BEN and BTB (POZ) domain containing; Nucleus accumbens-associated protein 2; Repressor with BTB domain and BEN domain; transcription repressor with a BTB domain and a BEN domain
Gene Aliases: 0610020I02Rik; AI448087; BEND9; BTBD14; BTBD14A; BTBD31; C030048H19Rik; NAC-2; NAC2; NACC2; RBB
UniProt ID: (Human) Q96BF6, (Mouse) Q9DCM7
Entrez Gene ID: (Human) 138151, (Mouse) 67991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support