Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC18120; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RING-type E3 NEDD8 transferase UBE2F; ubiquitin conjugating enzyme E2F (putative); Ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative)
Gene Aliases: 2510010F15Rik; AI851109; NCE2; RGD1307608; UBE2F; Ube2fa
UniProt ID: (Human) Q969M7, (Rat) Q5U203, (Mouse) Q9CY34
Entrez Gene ID: (Human) 140739, (Rat) 363284, (Mouse) 67921
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support