Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLRIPVRKAL VGLSKSPKGC VRTTATAASN LIEVFVDGQS VMVEPGTTVL QACEKVGMQI PRFCYHERLS VAGNCRMCLV EIEKAPKVVA ACAMPVMKGW NILTNSEKSK KAREGVMEFL LANHPLDCPI CDQGGECDLQ DQSMMFGNDR SRFLEGKRAV EDKNIGPLVK TIMTRCIQCT RCIRFASEIA GVDDLGTTGR GNDMQVGTYI EKMFMSELSG NIIDICPVGA LTSKPYAFTA QPWETRKTES IDVMDAVGSN IVVSTRTGEV MRILPRMHED INEEWISDKT RFAYDGLKRQ RLTEP
The protein encoded by this gene belongs to the complex I 75 kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-75kD; complex I 75kDa subunit; complex I, mitochondrial respiratory chain, 75-kD subunit; Complex I-75kD; MGC26839; mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase); NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial; NDUFS-1; OTTHUMP00000206345
Gene Aliases: 5830412M15Rik; 9930026A05Rik; CI-75k; CI-75Kd; NDUFS1; PRO1304
UniProt ID: (Human) P28331, (Rat) Q66HF1, (Mouse) Q91VD9
Entrez Gene ID: (Pig) 100516402, (Human) 4719, (Rabbit) 100341941, (Rat) 301458, (Mouse) 227197
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support