Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGYTKKVYEG GSNVDQFVTR FLLKETANQI QSLLSSVESA VEAIEEQTSQ LRQNHIKPSH SAAQTWCGSP TPASAPNHKL MAMEQGKTLP SATEDAKEEG LEAQISRLAE LIGRLESKAL WFDLQQRLSD EDGTNMHLQL VRQEMAVCPE QLSEFLDSLR QYLRGTTGVR NCFHITAVRL SDGFTFVIYE FWETEEAWKR HLQSPLCKAF RHVKVDTLSQ PEALSRILVP AAWCTVGRD
The protein encoded by this gene is a neuronal calcium-binding protein that binds to and modulates the function of at least two receptors, adenosine A(2A) receptor and metabotropic glutamate receptor type 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF hand calcium binding protein 2; EF-hand calcium binding protein 2; EF-hand calcium-binding protein 2; N-terminal EF-hand calcium-binding protein 2; neuronal calcium binding 2; Neuronal calcium-binding protein 2; Stip-2; Synaptotagmin-interacting protein 2
Gene Aliases: EFCBP2; NECAB2
UniProt ID: (Human) Q7Z6G3
Entrez Gene ID: (Human) 54550
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support