Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLRTESCRP RSPAGQVAAA SPLLLLLLLL AWCAGACRGA PILPQGLQPE QQLQLWNEID DTCSSFLSID SQPQASNALE ELCFMIMGML PKPQEQDEKD NTKRFLFHYS KTQKLGKSNV VSSVVHPLLQ LVPHLHERRM KRFRVDEEFQ SPFASQSRGY FLFRPRNGRR SAGFI (1-174 aa encoded by BC012908)
This gene encodes a member of the neuromedin family of neuropeptides. The encoded protein is a precursor that is proteolytically processed to generate a biologically active neuropeptide that plays a role in pain, stress, immune-mediated inflammatory diseases and feeding regulation. Increased expression of this gene was observed in renal, pancreatic and lung cancers. Alternative splicing results in multiple transcript variants encoding different isoforms. Some of these isoforms may undergo similar processing to generate the mature peptide.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuromedin-U; NmU 25; prepro-NMU
Gene Aliases: NMU
UniProt ID: (Human) P48645
Entrez Gene ID: (Human) 10874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support