Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PCNA-binding protein that acts as a regulator of DNA repair during DNA replication. Following DNA damage, the interaction with PCNA is disrupted, facilitating the interaction between monoubiquitinated PCNA and the translesion DNA synthesis DNA polymerase eta (POLH) at stalled replisomes, facilitating the bypass of replication-fork-blocking lesions. Also acts as a regulator of centrosome number.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HCV NS5A-transactivated protein 9; HCV NS5A-transactivated protein 9 homolog; Hepatitis C virus NS5A-transactivated protein 9; OEATC-1; Overexpressed in anaplastic thyroid carcinoma 1; p15PAF; PAF15; PCNA-associated factor; PCNA-associated factor of 15 kDa; PCNA-clamp-associated factor
Gene Aliases: AA409629; KIAA0101; L5; mKIAA0101; Ns5apt9; NS5ATP9; OEATC; OEATC-1; OEATC1; p15(PAF); p15/PAF; p15PAF; PAF; PAF15; PCLAF
UniProt ID: (Mouse) Q9CQX4, (Human) Q15004, (Rat) Q6RIA2
Entrez Gene ID: (Mouse) 68026, (Human) 9768, (Rat) 300795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support