Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chronic myelogenous leukemia tumor antigen 66; NudC domain-containing protein 1; NUDCD1; Tumor antigen CML66
Gene Aliases: 4921532K09Rik; AA407246; AW260430; AW556235; CML66; CML66-L; NUDCD1; OVA66; RGD1310624
UniProt ID: (Human) Q96RS6, (Mouse) Q6PIP5
Entrez Gene ID: (Human) 84955, (Rat) 362906, (Mouse) 67429
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support