Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHAPGMKKIK LDTPEEIARW REERRKNYPT LANIERKKKL KLEKEKRGAV LTTTQYGKMK GMSRHSQMAK IRSPGKNHKW KNDNSRQRAV TGSGSHLCDL KLEGPPEANA DPLGVLINSD SESDKEEKPQ HSVIPKEVTP ALCSLMSSYG SLSGSESEPE ETPIKTEADV LAENQVLDSS APKSPSQDVK ATVRNFSEAK SENRKKSFEK TNPKRKKDYH NYQTLFEPRT HHPYLLEMLL APDIRHERNV ILQCVRYIIK KDFFGLDTNS AKSKDV
This gene encodes a nuclear RNA binding protein that contains a C2H2 zinc finger motif and a nuclear localization signal. This protein is associated with the nuclear matrix in perichromatin fibrils and, in neurons, localizes to the cytoplasm in association with endoplasmic reticulum ribosomes. This protein interacts with the fragile X mental retardation protein (FMRP), the tumor suppressor protein BRCA1, upregulates RNA polymerase II transcription, and is involved in box C/D snoRNP biogenesis. A pseudogene of this gene resides on chromosome 6q12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FMR1-interacting protein NUFIP1; Nuclear FMR1-interacting protein 1; Nuclear FMRP-interacting protein 1; nuclear fragile X mental retardation protein interacting protein 1
Gene Aliases: bA540M5.1; NUFIP; NUFIP1
UniProt ID: (Human) Q9UHK0
Entrez Gene ID: (Human) 26747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support