Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVWGHYDLC KTQIYTEEGK VWDYMACQPE STDMTKYLKV KLDPPDITCG DPPETFCAMG NPYMCNNECD ASTPELAHPP ELMFDFEGRH PSTFWQSATW KEYPKPLQVN ITLSWSKTIE LTDNIVITFE SGRPDQMILE KSLDYGRTWQ PYQYYATDCL DAFHMDPKSV KDLSQHTVLE IICTEEYSTG YTTNSKIIHF EIKDRFAFFA GPRLRNMASL YGQLDTTKKL RDFFTVTDLR IRLLRPAVGE IFVDELHLAR YFYAISDIKV RGRCKCNLHA TVCVYDNSKL TCECEHNTTG PDCGKCKKNY QGRPWSPGSY LPIPKGTANT CIPSISSIGT (25-363 aa encoded by BC030220)
Netrin G1 (NTNG1) belongs to a conserved family of proteins that act as axon guidance cues during vertebrate nervous system development (Nakashiba et al., 2000).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: axon guidance molecule; laminet 1; Laminet-1; netrin G1f; Netrin-G1; YLSR571
Gene Aliases: A930010C08Rik; AI853992; KIAA0976; LMNT1; NTNG1; RGD1563465; UNQ571/PRO1133
UniProt ID: (Human) Q9Y2I2, (Mouse) Q8R4G0
Entrez Gene ID: (Human) 22854, (Rat) 295382, (Mouse) 80883
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support