Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Neuromedin U (NmU) is a vertebrate peptide which stimulates uterine smooth muscle contraction and causes selective vasoconstriction. Like most other active peptides, it is proteolytically processed from a larger precursor protein. The mature peptides are 8 (NmU-8) to 25 (NmU-25) residues long and C-terminally amidated. The sequence of the C-terminal extremity of NmU is extremely well conserved across species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuromedin-U; NmU 25; prepro-NMU
Gene Aliases: NMU
UniProt ID: (Human) P48645, (Rat) P12760, (Mouse) Q9QXK8
Entrez Gene ID: (Human) 10874, (Rat) 63887, (Mouse) 56183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support